Sulfur Standard: S @ 5 ug/g in 80% Isooctane/20% Toluene, VHG, LGC Standards
Catalog # 77695-543
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:5 ug
- Description:Sulfur Standard: S @ 5 ug/g in 80% Isooctane/20% Toluene
- Size:100 ml
- Cat. No.:77695-543