Sulfur Standard: S @ 1000 µg/g (0.100 wt%) in Kerosene, VHG, LGC Standards
Catalog # 77695-546
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:1000 µg
- Description:Sulfur Standard: S @ 1000 µg/g (0.100 wt%) in Kerosene
- Size:100 ml
- Cat. No.:77695-546