Multi-Element Standard Alkalis and Alkaline Earths 10 Components: 100 µg/mL each of Ba, Be, Ca, Cs, K, Li, Mg, Na, Rb, Sr in 5% HNO3, VHG, LGC Standards
77693-257
New Product
:
- Concentration:100 µg/mL
- Matrix:5% HNO₃
- Description:Alkalis & Alkaline Earths Standard: Ba, Be, Ca, Cs, K, Li, Mg, Na, Rb, Sr @ 100 µg/mL in 5% HNO3
- Size:100 ml
- Cat. No.:77693-257