Order Entry
Puerto Rico
ContactUsLinkComponent
Sulfur Standard: S @ 10 µg/g (0.0010 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Sulfur Standard: S @ 10 µg/g (0.0010 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-562
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Sulfur Standard: S @ 10 µg/g (0.0010 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-562
Supplier:  LGC Standards
Supplier Number:  VHG-SMIN-10-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10 µg/g
  • Matrix:
    Mineral oil
  • Description:
    Sulfur Standard: S @ 10 µg/g (0.0010 wt%) in 75 cSt Mineral Oil
  • Size:
    100 ml
  • Cat. No.:
    77695-562

Specifications

About this item