Sulfur Standard: S @ 1000 µg/g (0.100 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
77695-560
New Product
:
- Concentration:1000 µg/g
- Matrix:Mineral oil
- Description:Sulfur Standard: S @ 1000 µg/g (0.100 wt%) in 75 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-560