Sulfur Standard: S @ 500 µg/g (0.0500 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-578
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:500 µg/g
- Matrix:Mineral oil
- Description:Sulfur Standard: S @ 500 µg/g (0.0500 wt%) in 75 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-578