Sulfur Standard: S @ 5000 µg/g (0.500 wt%) in 75 cSt Mineral Oil, VHG, LGC Standards
77695-577
New Product
:
- Concentration:5000 µg/g
- Matrix:Mineral oil
- Description:Sulfur Standard: S @ 5000 µg/g (0.500 wt%) in 75 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-577