Multi-Element Three Component Standard: 5.0 wt% S, 100 µg/g Fe, 50 µg/g V in 20 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-588
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Mineral oil
- Description:Sulfur & Metals in Oil Standard 12: S @ 5.0 wt%, Fe @ 100 µg/g, Ni @ 0 µg/g, V @ 50 µg/g in 20 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-588