Multi-Element Four Component Standard: 4.5 wt% S, 250 µg/g Fe, 60 µg/g Ni, 100 µg/g V in 20 cSt Mineral Oil, VHG, LGC Standards
Catalog # 77695-591
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Mineral oil
- Description:Sulfur & Metals in Oil Standard 5: S @ 4.5 wt%, Fe @ 250 µg/g, Ni @ 60 µg/g, V @ 100 µg/g in 20 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-591