Multi-Element Four Component Standard: 5.5 wt% S, 50 µg/g, Fe, 40 µg/g Ni, 400 µg/g V in 20 cSt Mineral Oil, VHG, LGC Standards
77695-594
New Product
:
- Matrix:Mineral oil
- Description:Sulfur & Metals in Oil Standard 8: S @ 5.5 wt%, Fe @ 50 µg/g, Ni @ 40 µg/g, V @ 400 µg/g in 20 cSt Mineral Oil
- Size:100 ml
- Cat. No.:77695-594