Sulfur Standard: S @ 50000 µg/g (5.00 wt%) in Residual Oil, VHG, LGC Standards
Catalog # 77695-612
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Concentration:50000 µg/g
- Matrix:oil
- Description:Sulfur Standard: S @ 50000 µg/g (5.00 wt%) in Residual Oil
- Size:100 ml
- Cat. No.:77695-612