Order Entry
Puerto Rico
ContactUsLinkComponent
Tantalum Standard: Ta @ 10000 µg/mL in 2% HF, VHG, LGC Standards
Tantalum Standard: Ta @ 10000 µg/mL in 2% HF, VHG, LGC Standards
Catalog # 77695-626
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Tantalum Standard: Ta @ 10000 µg/mL in 2% HF, VHG, LGC Standards
Catalog # 77695-626
Supplier:  LGC Standards
Supplier Number:  VHG-TTAF-100
New Product
CAS Number:  

Specifications

  • Concentration:
    10000 µg/mL
  • Matrix:
    2% HF
  • Description:
    Tantalum Standard: Ta @ 10000 µg/mL in 2% HF
  • Size:
    100 ml
  • Cat. No.:
    77695-626

Specifications

About this item