Multi-Element Standard 23 components: 300 µg/g each of Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG, LGC Standards
Supplier: LGC Standards
New Product
Certificates
About this item
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.