Tacta® Mechanical Pipettes
Supplier: Sartorius
Certificates
About this item
Tacta is a premium mechanical pipette with ergonomic design for comfortable pipetting and reliability.
- Low pipetting cycle forces reduce the risk of strain injury
- Optiload feature in both single and multichannel models for easy and light tip loading with perfect tip sealing
- Optiject soft tip ejection feature detaches the tip in a smooth, controlled manner
- Optilock’s dual-function volume locking prevents accidental volume changes
- Easy-to-read, four-digit volume display helps to set exact volumes
- Color-coding of volumes to ease the selection of corresponding pipette tips
- Safe-Cone Filters available for models above 10 μl with convenient built-in filter ejection mechanism
Designed with comfort in mind, Tacta makes pipetting effortless and safe, while producing accurate and reliable results time after time. Tacta is made from carefully selected materials, with each component designed to meet the highest standards.
As Tacta was designed with comfort in mind, it fits both in your hand and in your laboratory. As it requires less force than other mechanical pipettes on the market, it can be used for longer pipetting sessions without causing repetitive strain injuries, or affecting the reproducability and reliability of the results.
These pipettes have no battery and are fully autoclavable without disassembling.