Human/Mouse Recombinant TGF-β 3 (Animal free) (from E. coli)
Supplier: Fujifilm Irvine Scientific
New Product
Certificates
About this item
Transforming growth factors (TGFs) are multifunctional peptides that regulate growth and differentiation in most cell types. The TGF-β family of proteins signal through serine/threonine kinase receptors. TGF-β isoforms (TGF-β1, -β2, and –β3) have overlapping, yet distinct biological actions in developing and adult tissues. TGF-β3 is an important factor in regulating cell adhesion and accelerating wound repair. TGF-β3 also functions during osteoblast proliferation, chemotaxis, and collagen synthesis.
- High Quality
- Low Endotoxin
- Affordable
Shelf life: 12 months from date of receipt when stored at 4 °C as supplied.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:HumanMouse
- Storage Conditions:Store at 4 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: Inhibition of IL-4-induced HT-2 cell proliferation, ED50 ≤1 ng/ml, Bioactivity ≥1.0×10⁶ units/mg
- Form:Liquid
- Gene ID:P10600/P17125
- Amino Acid Number:113/226
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein/Peptide Name:TGF-β 3
- Purity:≥95%
- Molecular Weight:12.9/25.7 kDa
- Sequence:MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
- Endotoxin Level:Low
- Formulation:10 mM acetic acid and 20% ethanol at a concentration of 0.25 mg/ml
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:Ice pack
- Tested Applications:Bioassay
- Purification:Reducing and non-reducing SDS-PAGE analysis
Specifications
Frequently Bought Together