Human Recombinant FGF-9 (Animal free) (from E. coli)
Supplier: Fujifilm Irvine Scientific
Certificates
About this item
Fibroblast growth factor 9 (FGF-9) is a mitogen and survival factor for nerve and mesenchymal cells. FGF-9 functions as an autocrine and paracrine factor to support the growth and survival of motor neurons and prostate tissue. FGF-9 expression in the gonad is also necessary for sex determination.
- High Quality
- Low Endotoxin
- Affordable
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Endotoxin LAL (EU/µg) ≤0.1
- Biological Activity:Bioactivity assay: NR6R 3T3 cell proliferation, ED50 ≤2 ng/ml, Bioactivity ≥5.0×10⁵ units/mg
- Gene ID:P31371
- Reconstitution Instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Heparin-binding growth factor -9 (HBGF-9)|Glia activating factor (GAF)
- Protein/Peptide Name:RH FGF-9 (AF)
- Purity:≥95%
- Molecular Weight:23.4 kDa
- Sequence:MPLGEVGNYF GVQDAVPFGN VPVLPVDSPV LLSDHLGQSE AGGLPRGPAV TDLDHLKGIL RRRQLYCRTG FHLEIFPNGT IQGTRKDHSR FGILEFISIA VGLVSIRGVD SGLYLGMNEK GELYGSEKLT QECVFREQFE ENWYNTYSSN LYKHVDTGRR YYVALNKDGT PREGTRTKRH QKFTHFLPRP VDPDKVPELY KDILSQS
- Endotoxin Level:Low
- Formulation:10 mM sodium phosphate, 25 mM sodium chloride, 50 mM sodium sulfate, pH 7.5
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Tested Applications:Bioassay
Specifications
Frequently Bought Together