Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ala-Phe

Ala-Phe

Supplier:   Aladdin Scientific
Ala-Phe
Supplier:  Aladdin Scientific
Certificates
Ala-Phe

3 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
  • GHS Signal word: Warning
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ALA121378250MG
  • ALA121378-1G
  • ALA121378-5G
  • Protein/Peptide Name:
  • Ala-Phe
  • Ala-Phe
  • Ala-Phe
  • Protein Synonyms:
  • NSC 89630|L-Alanyl-L-phenylalanine|Alanylphenylalanine|L-Ala-L-phe
  • NSC 89630|L-Alanyl-L-phenylalanine|Alanylphenylalanine|L-Ala-L-phe
  • NSC 89630|L-Alanyl-L-phenylalanine|Alanylphenylalanine|L-Ala-L-phe
  • CAS No.:
  • 3061-90-3
  • 3061-90-3
  • 3061-90-3
  • MDL No.:
  • MFCD00066031
  • MFCD00066031
  • MFCD00066031
  • Purity:
  • ≥98%
  • ≥98%
  • ≥98%
  • Molecular Weight:
  • 236.27 g/mol
  • 236.27 g/mol
  • 236.27 g/mol
  • Molecular Formula:
  • C₁₂H₁₆N₂O₃
  • C₁₂H₁₆N₂O₃
  • C₁₂H₁₆N₂O₃
  • Storage Conditions:
  • Freezer
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Glass bottle
    250 mg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $18.90
    $18.90
    Each(1/Itm/Bx)
       $18.90

    Supplier Number: A121378-250MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    ALA121378250MG
    Protein/Peptide Name
    Ala-Phe
    Protein Synonyms
    NSC 89630|L-Alanyl-L-phenylalanine|Alanylphenylalanine|L-Ala-L-phe
    CAS No.
    3061-90-3
    MDL No.
    MFCD00066031
    Purity
    ≥98%
    Molecular Weight
    236.27 g/mol
    Molecular Formula
    C₁₂H₁₆N₂O₃
    Storage Conditions
    Freezer
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $57.90
    $57.90
    Each(1/Itm/Bx)
       $57.90

    Supplier Number: A121378-1G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    ALA121378-1G
    Protein/Peptide Name
    Ala-Phe
    Protein Synonyms
    NSC 89630|L-Alanyl-L-phenylalanine|Alanylphenylalanine|L-Ala-L-phe
    CAS No.
    3061-90-3
    MDL No.
    MFCD00066031
    Purity
    ≥98%
    Molecular Weight
    236.27 g/mol
    Molecular Formula
    C₁₂H₁₆N₂O₃
    Storage Conditions
    Freezer
  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $228.00
    $228.00
    Each(1/Itm/Bx)
       $228.00

    Supplier Number: A121378-5G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    ALA121378-5G
    Protein/Peptide Name
    Ala-Phe
    Protein Synonyms
    NSC 89630|L-Alanyl-L-phenylalanine|Alanylphenylalanine|L-Ala-L-phe
    CAS No.
    3061-90-3
    MDL No.
    MFCD00066031
    Purity
    ≥98%
    Molecular Weight
    236.27 g/mol
    Molecular Formula
    C₁₂H₁₆N₂O₃
    Storage Conditions
    Freezer

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.