About this item
Designed to improve the accuracy of PCR and general tissue culture procedures
The clear, replaceable 13mm (½") thick front viewing panel is effective against beta rays and 32P labeled compounds. Hand knobs on hinged corner doors allow access to the chamber. Electrical sensors turn off power to UV system when door is opened.
Unit significantly reduces the risk of airborne contamination during DNA sequencing. Still air enclosure chamber contains both fluorescent and UV germicidal lamps. This UV system is rated at 254nm and decontaminates all interior surfaces. Lamps should be active 15 minutes before and after use for proper decontamination. Chamber with HEPA filter features a positive pressure external blower rated at <40dBA and a small HEPA filter rated at 99.7% efficiency at 0.3µm.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 97000-992
- 10718-050
- Electrical:
- 110V, 60Hz
- 110V, 60Hz