- Description:Celloger® attachment for biochip, triple
- Cat. No.:77663-858
- For use with:Glass slides
With Celloger®, cells cultured in different vessel types can be imaged. Interchangeable vessel adapters can accommodate various vessel types including well-plate, culture flask, and culture dish.
The system includes a well-plate adapter, but you can buy separately different adapters for a variety of vessels used.