Orders LinkContactUsLinkComponent
Electrode Kit USC
Electrode Kit USC
Electrode kit, 2 V, 2 A
Catalog # 77664-238
Supplier #:  EK1
Electrode Kit USC
Catalog # 77664-238
Electrode Kit USC
Electrode kit, 2 V, 2 A
Price
$499.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The electrode kit contains four voltage/current electrodes, plus four luer-tipped cartridges. Electrodes are threaded and screw securely into the end of each cartridge.
The luer tip then plugs securely into the luer openings of the chamber. The cable from each electrode terminates with a 2 mm pin which may be plugged into voltage/current clamps such as WPI’s DVC1000 or EVC-4000.
The miniature electrode-gel cartridge is a small plastic tube with a male luer tip identical to those at the tip of hypodermic syringes. The tube may be filled with different gel materials; agar is commonly used but other gel materials may also be satisfactory.
Specifications
  • Description:
    Electrode kit, 2 V, 2 A