Orders LinkContactUsLinkComponent
EVOM™ Auto Wired Connection Kit
EVOM™ Auto Wired Connection Kit
EVOM™ auto wired connection kit
Catalog # 77664-266
Supplier #:  EVA-AC-03-01
EVOM™ Auto Wired Connection Kit
Catalog # 77664-266
EVOM™ Auto Wired Connection Kit
EVOM™ Auto Wired Connection Kit
EVOM™ auto wired connection kit
Price
$95.00
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit is for peer-to peer wired connection set up.
The set up requires EVOM™ Auto system’s Software rev 3.0.0.
Delivery information: This kit contains:
(1) USB 3.0 GB ethernet adapter
(1) Ethernet cable 25'
(1) EVOM™ auto ethernet update USB flash drive
Specifications
  • Description:
    EVOM™ auto wired connection kit