ContactUsLinkComponent
Omega-Tip-Z™ Electrode Impedance Meter
Omega-Tip-Z™ Electrode Impedance Meter
Catalog # 77665-192
New Product
Omega-Tip-Z™ Electrode Impedance Meter
Catalog # 77665-192
Supplier Number:  SYS-OMEGAZ
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Electrode impedance meter
  • Cat. No.:
    77665-192
Specifications
About this item
Omega-Tip-Z™ was created especially for measuring impedance in etched tung­sten, platinum-iridium* and steel microelectrodes, as well as electrolyte-filled micropipettes. The meter's AC impedance-measuring circuit is unaffected by electrode offset or tip junction potentials.
  • Battery operated
  • Portable
  • Measures mV and MΩ impedance of metal or glass microelectrodes
  • Low noise probe included
  • Determine resistance of glass capillary electrode during micropipette beveling process.
  • Measure impedance of metal microelectrodes
The gold-plated miniature probe lets you conveniently monitor micro-electrode impedance in electrolytes, and an electrode tip cleaning feature lets you remove buildup quickly. Omega-Tip-Z can also measure DC electrode tip potentials up to 2000 millivolts. The instrument operates for hundreds of hours without battery failure.
Applications: Impedance measurements provide valuable information about the electrode-electrolyte interface, including the resistance and capacitance of the system.