Nalgene® Staining Boxes, Thermo Scientific
Catalog # 28196-310
Supplier: Thermo Fisher Scientific
Specifications
- Description:Staining Boxes
- Cat. No.:28196-310
- Interior dimensions:12.7 x 12.7 x 5.1 cm (5 x 5 x 2")
- Supplier no.:5705-1010
Specifications
About this item
Boxes are made of polymethylpentene and feature a polypropylene copolymer cover and a low-density polyethylene plug
Use box to stain, fix, destain, and handle electrophoresis gels and membranes with less chance of damage than conventional methods. Box 28196-310 holds up to 500mL of liquid when a gel is in place; box 28196-306 holds up to 750mL. Both box and cover will resist temperatures from –70 to 70°C (–94 to 158°F), and have good chemical resistance to acids, bases, and select organic solvents. Strategic plug location allows easy liquid drainage with minimal gel handling. The plug will not interfere with the cover or box contents.