Orders LinkContactUsLinkComponent
Filter Pumps, Richards
Filter Pumps, Richards
Filter Pumps, Richards
Certificates
Filter Pumps, Richards
Filter Pumps, Richards
1 Options Available Below
Product Details & Documents
Z-form tail design causes violent turbulence with consequent thorough mixture of water and entrained air
It is impossible for the air to re-enter the suction tube. Provided with male threads on all three connectors.
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 28640-020
    Filter pump 3/8" IPS thread
    Filter pump 3/8" IPS thread
    7"
    Discontinued
    Supplier Number: H-12120
  • 28640-042
    Filter pump 3/4" IPS thread
    Filter pump 3/4" IPS thread
    13"
    Log in for availability
    Log in for availability
    Each
    Each - $440.56
    $440.56
    Each
       $440.56
    Supplier Number: H-12130