Weller® Cordless Soldering Iron Kit, Cooper Tools, Apex
Catalog # 37698-362
Supplier: Cooper Tools
Specifications
- Description:Cordless Soldering Iron Kit
- Cat. No.:37698-362
- Supplier no.:PSI100K
Specifications
About this item
Professional-quality Portasol® heat tool to solder, heat shrink, and cut rope
Butane fuel level is visible through a window on the body of the tool. Built-in fuel filter reduces fuel contamination. Cap includes auto safety shut-off switch. Operates up to two hours per refill. Temperature is adjustable up to 580°C (1076°F). Kit contains iron, sponge, stand, and extra tips, all packed in a plastic case.