ContactUsLinkComponent
SuprClean™ Defluxer, MicroCare
SuprClean™ Defluxer, MicroCare
Catalog # 76020-174
Supplier:  MicroCare
undefined
SuprClean™ Defluxer, MicroCare
Catalog # 76020-174
Supplier:  MicroCare
Supplier Number:  MCC-SPR
Specifications
  • Size:
    340 g (12 oz.)
  • Environmentally Preferable:
  • Cat. No.:
    76020-174
Specifications
About this item
An extremely aggressive HFC-based solvent for fast, easy cleaning
Great defluxer, especially for rosin fluxes. Degreaser for all types of organic contamination. Fast-drying and nonflammable with good toxicity ratings. Ozone-safe. No aroma. Test on soft plastics. Aerosol spray can.