Flux-Off® Heavy-Duty Flux Remover, ITW Chemtronics®
22221-830
- Size:340 g (12 oz.)
- Environmentally Preferable:
- Cat. No.:22221-830
- Supplier no.:ES1631
For the removal of all rosin and encrusted no-clean flux types from electronic assemblies, printed circuit boards, and all other electronic components
Quick-drying flux remover is nonflammable, leaves no residue, and is ozone-safe. CFC/HCFC-free. Aerosol spray.