Reaction Flask, [ST] 24/40 Joint, Wheaton
Catalog # 80093-772
Supplier: DWK Life Sciences (KIMBLE)
Small Business Enterprise
Specifications
- Capacity:200 ml
- Description:Reaction Flask
- Cat. No.:80093-772
- Supplier no.:635965
Specifications
About this item
This flask is constructed of Wheaton 33® low extractable borosilicate glass
The design of the flask eliminates the ball and socket joint connecting the scrubber and absorber components used during the silver diethyldithiocarbamate method described in the 18th Edition of Standard Methods for the Examination of Water and Wastewater. Flask meets ASTM Type I Class A and USP Type I standards.