ContactUsLinkComponent
PVK Series Isolation Vacuum Valve, Edwards
Catalog # EVC311-05-000
Supplier:  Edwards
undefined
PVK Series Isolation Vacuum Valve, Edwards
Catalog # EVC311-05-000
Supplier:  Edwards
Supplier Number:  C31105000
Specifications
  • Description:
    Isolation Vacuum Valve
  • Cat. No.:
    EVC311-05-000
  • Supplier no.:
    C311-05-000
Specifications
About this item
Clean, bellows valve for manual operation.
Quick-acting, lever operated. The lever, connected to a self-lubricating plastic cam-piston, actuates the valve stem and valve plate through PTFE bearings. This mechanism is isolated from the vacuum system by a laser-welded, stainless steel bellows with minimum life expectancy of 1,000,000 operations. Valve is suitable for vacuum range of atmosphere to 10-9 torr. NW10 flange size.