Mini Dissecting Scissors, Left Hand
About this item
Our Left-Hand Mini Dissecting Scissors are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces.
- Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries
- Have thinner (and often longer) blades than operating scissors
- Designed for cutting and probing in tight spaces
- Designed for left handed people
They are commonly used in microsurgery, like ophthalmic surgery. These scissors are 9 cm (3.5") long and have straight blades with sharp tips or curved blades with fine tips.
These are available in surgical grade stainless steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
2 Options Available Below
Product Details & Documents
Our Left-Hand Mini Dissecting Scissors are similar to regular dissecting scissors, but they are shorter and have more delicate tips, making them ideal for fine dissection work in tight spaces.
- Our dissecting scissors are for general use in dissection, preparation of tissue samples, and small animal surgeries
- Have thinner (and often longer) blades than operating scissors
- Designed for cutting and probing in tight spaces
- Designed for left handed people
They are commonly used in microsurgery, like ophthalmic surgery. These scissors are 9 cm (3.5") long and have straight blades with sharp tips or curved blades with fine tips.
These are available in surgical grade stainless steel. Our Dissecting Scissors are designed to withstand the high heat and chemical exposure of the sterilization process. They may be autoclaved. To guarantee a long life for your surgical instruments, be sure to clean them thoroughly after each use.
Product Family Options
Product Information
- LengthMaterialCurvatureAvailabilityPrice
- 77663-338Mini dissecting scissor, fine tipsMini dissecting scissor, fine tips9 cm (3.5")Stainless steelCurvedLog in for availabilityEach$63.15Supplier Number: 503627
- 77663-336Mini dissecting scissor, sharp tipsMini dissecting scissor, sharp tips9 cm (3.5")Stainless steelStraightLog in for availabilityEach$63.15Supplier Number: 503625