Specifications
- Description:Luer valve assortment kit
- Cat. No.:77661-658
Specifications
About this item
This Luer fittings kit is useful for building your own liquid flow experiment. It provides the means to start, stop, add, divide and control a flow of liquid or gas.
- Over 300 assorted Luer fittings and parts
- Luer fittings for quick and easy connect and disconnect
- Applications: Liquid flow experimental setups
Included in the kit are over 300 assorted parts such as one-way and 3-way stopcocks, manifolds, Y-connectors, injection sites, male and female luer caps, check valves, syringe-activated check valves, slide clamps, roller clamps, and pinch clamps. All (except clamps) have a luer fitting for quick and easy connecting and disconnecting. This Luer fittings kit ncludes assorted luer fittings for use with flexible tubing.