Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Calcium Buffer Kit

Calcium Buffer Kit

Calcium buffers kit, 8 bottles, 20 ml
Catalog # 77664-080
Supplier #:  CALBUF-1
Calcium Buffer Kit
Catalog # 77664-080
CAS Number:  
Calcium Buffer Kit

Calcium buffers kit, 8 bottles, 20 ml

CAS Number:  
Price
$180.00
Log in for availability
Pack of 8 (20ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

A set of eight calcium buffers covering the range of concentration from 10⁻¹ to 10⁻⁸ M Ca⁺⁺. Each buffer contains 20 ml of solution and enough potassium chloride to set the ionic strength to 0.1 M.

  • For use with calcium electrodes
  • 20 ml of each buffer

Concentrations: 1×10⁻¹, 1×10⁻², 1×10⁻³, 1×10⁻⁴, 1×10⁻⁵, 1×10⁻⁶, 1×10⁻⁷, 1×10⁻⁸ M at 20 °C.

Specifications

Specifications

  • Pack type:
    8 bottles
  • Description:
    Calcium buffers kit
  • Size:
    20 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.