ContactUsLinkComponent
Mouse Rectal Probe Ball Tip
Mouse Rectal Probe Ball Tip
Catalog # 77663-918
New Product
Mouse Rectal Probe Ball Tip
Catalog # 77663-918
Supplier Number:  61824
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Mouse rectal probe ball tip
  • Length:
    1.1 m
  • Cat. No.:
    77663-918
Specifications
About this item
The needle microprobes and animal rectal probes are all designed to withstand sterilization by ETO gas and gluteraldehyde solution. Our flexible implantable microprobes are both ETO gas sterilizable and autoclavable.
  • Rectal probe for use with the ATC2000, Animal temperature controller
  • The heating plate cable is 1.1 m long