Orders LinkContactUsLinkComponent
Botanical Kit
Dissecting kit
  77662-618
Botanical Kit
  77662-618
Botanical Kit
Dissecting kit
$38.51
Each
Basic kit for examining biological specimens studies.
  • Contains 9 pieces
 : Kit inclues: Dissecting scissors, 4.5", dissecting scissors, 4"Dressing forceps, 5.5", micro dressing forceps, 4", #4 knife handle, straight teasing needle, angled teasing needle, #22 surgical blade, canvas roll-up.
  • Description:
    Dissecting kit