- Pack type:Vial
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Mouse
- Environmentally Preferable:
- Storage Conditions:At or below −20 °C
- Biological Activity:Bioactivity: No biological activity data is available at this time
- Endotoxin-free:N
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:N
- Protein/Peptide Name:MOUSE HSC EXPANSION PB
- Endotoxin Level:Low
- Nuclease-free:N
- Grade:RUO
- Shipping Temperature:RT
- Cat. No.:77661-208
Cytokines for polarizing T cells into T regulatory cells. T regulatory cells (Tregs) are a subset of T cells that suppress immune function. They are characterized by the expression of CD4, CD25 and Foxp3 and are known to produce immunosuppressive cytokines TGF-β 1 and IL-10. Tregs can be cultured from naive CD4+ T cells by culturing them in the presence of IL-2 and TGF-β 1 with CD3 and CD28 stimulation. In some cases cells are also cultured with neutralizing antibodies to IL-4, IFNγ and IL-12 and/or retinoic acid.
- High quality
- Low endotoxin
- Affordable
Studies have shown that decreased expression of CD34 tends to correlate with decreased self renewal properties and pluripotent potential. Culturing CD34+ cells in a cytokine mixture of TPO, SCF, and FLT-3 Ligand for short time periods allows CD34+ cells to proliferate without decreasing CD34 expression, thus retaining their stem cell properties.