Lithium tetraborate : lithium metaborate : lithium bromide (35.05% : 64.45% : 0.5%), granular reference standard, VHG
Catalog # 77693-337
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 kg
- Cat. No.:77693-337
Specifications
About this item
: