Water Pollution QC Check Sample: Oil and Grease Concentrate, VHG, LGC Standards
Catalog # 77695-432
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:n-Propanol
- Pack type:Glass bottle
- Description:Water Pollution QC Check Sample: Oil and Grease Concentrate
- Size:23 ml
- Cat. No.:77695-432