VOC Mixture 659 2000 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-384
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- Pack type:Glass bottle
- Description:VOC Mi xture 659 2000 µg/mL in Methanol
- Size:1 ml
- Cat. No.:77677-384