Sulfonamides Mixture for GB/T 21316-2007 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77693-479
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Acetonitrile
- Pack type:Glass bottle
- Description:Sulfonamides Mi xture for GB/T 21316-2007 100 µg/mL in Acetonitrile
- Size:1 ml
- Cat. No.:77693-479