Pesticide-Mix 20 10 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-139
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Cyclohe xane
- Pack type:Glass bottle
- Description:Pesticide-Mi x 20 10 µg/mL in Cyclohe xane
- Size:10 ml
- Cat. No.:77679-139