Pesticide-Mix 192 5-10 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-146
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Isooctane
- Pack type:Glass bottle
- Description:Pesticide-Mi x 192 5-10 µg/mL in Isooctane
- Size:10 ml
- Cat. No.:77679-146