LC PestiMix 2 5 µg/mL in Acetonitrile:Acetone (94.5:5.5), Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-329
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Acetonitrile:Acetone
- Pack type:Glass bottle
- Description:LC PestiMi x 2 5 µg/mL in Acetonitrile:Acetone (94.5:5.5)
- Size:1 ml
- Cat. No.:77677-329