GC PestiMix 4 10 µg/mL in Isooctane:Toluene (50:50), Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-352
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Isooctane:Toluene
- Pack type:Glass bottle
- Description:GC PestiMi x 4 10 µg/mL in Isooctane:Toluene (50:50)
- Size:1 ml
- Cat. No.:77677-352