EPA Method 625/8270 Composites Mixture 2000 µg/mL in Dichloromethane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-347
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Dichloromethane
- Pack type:Glass bottle
- Description:EPA Method 625/8270 Composites Mi xture 2000 µg/mL in Dichloromethane
- Size:1 ml
- Cat. No.:77677-347