Dichloromethane 100 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-892
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Methanol
- Pack type:Glass bottle
- Description:Dichloromethane 100 µg/mL in Methanol
- Size:1 ml
- Cat. No.:77691-892