California Residual Solvent Mixture 2B 1000 µg/mL in Dimethyl Sulfoxide Second Source, Dr. Ehrenstorfer, LGC Standards
Catalog # 77677-358
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- Matrix:Dimethyl Sulfo xide
- Pack type:Glass bottle
- Description:California Residual Solvent Mi xture 2B 1000 µg/mL in Dimethyl Sulfo xide Second Source
- Size:1 ml
- Cat. No.:77677-358