$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Ziprasidone-d8, TRC, LGC Standards
Ziprasidone-d8, TRC, LGC Standards
Ziprasidone-d8, TRC, LGC Standards
Catalog # 77695-306
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Ziprasidone-d8, TRC, LGC Standards
Catalog # 77695-306
Supplier:
LGC Standards
Supplier Number:
TRC-Z485002-1MG
New Product
CAS Number:
Specifications
CAS No.:
1126745-58-1
Description:
Ziprasidone-d8
Size:
1 mg
Cat. No.:
77695-306
Specifications
About this item
Product Details
Product Details