$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Vitamin B12 e-Monocarboxylic Acid, TRC, LGC Standards
Vitamin B12 e-Monocarboxylic Acid, TRC, LGC Standards
77695-290
:
LGC Standards
New Product
:
Vitamin B12 e-Monocarboxylic Acid, TRC, LGC Standards
77695-290
:
LGC Standards
:
TRC-V675655-10MG
New Product
:
CAS No.:
26264-28-8
Description:
Vitamin B12 e-Monocarboxylic Acid
Size:
10 mg
Cat. No.:
77695-290