Troleandomycin, TRC, LGC Standards
Catalog # 77695-268
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2751-09-9
- Description:Troleandomycin
- Size:2.5 mg
- Formula:C₄₁H₆₇NO₁₅
- Molecular Weight:814 g/mol
- Cat. No.:77695-268