Tris(2,4-di-tert-butylphenyl)phosphate, TRC, LGC Standards
Catalog # 77695-265
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:95906-11-9
- Description:Tris(2,4-di-tert-butylphenyl)phosphate
- Size:100 mg
- Formula:C₄₂H₆₃O₄P
- Molecular Weight:662.92 g/mol
- Cat. No.:77695-265